Learn More
Medchemexpress LLC HY-N0480A 100mg Medchemexpress, Reserpine (hydrochloride) CAS:16994-56-2 Purity:>98%
SDP
Catalog No. 501933593
Encompass_Preferred
Medchemexpress, HY-N0480A 100mg Reserpine (hydrochloride) CAS:16994-56-2 Reserpine hydrochloride is an inhibitor of the vesicular monoamine transporter 2 ( VMAT2 ). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3593 Supplier Medchemexpress LLC Supplier No. HYN0480A100MG
Medchemexpress, HY-N0480A 100mg Reserpine (hydrochloride) CAS:16994-56-2 Reserpine hydrochloride is an inhibitor of the vesicular monoamine transporter 2 ( VMAT2 ). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.