Learn More
Waters Corp Reserpine Stock and Working Solution
Supplier:  Waters Corp 700001673
Encompass_Preferred
Standards used for Mass Spec. Reserpine Stock Solution 100pg/ul, +1%,-0% in 50/50 Methanol Water 15mL Nalgene Brown Bottle Reserpine Working Solution, 1pg/ul, +1%,-0% in 50/50 Methanol/Water, 15 mL Brown Nalgene
Catalog No. 50-201-660