Learn More
Enzo Life Sciences Resveratrol, (100mg), CAS Number: 501-36-0
SDP
Supplier:  Enzo Life Sciences BMLFR1040100
Encompass_Preferred
Antioxidant. SIRT1 activator. Alternative Name: trans-3,4',5-Trihydroxystilbene. Formula: C14H12O3. MW: 228.2. CAS Number: 501-36-0. MI: 14: 8158. Purity: ≥98% (HPLC). Appearance: White to yellow solid. Solubility: Soluble in DMSO (65mg/ml), 100% ethanol or dimethyl formamide; slightly soluble in water (0.1mg/ml). Use/Stability: Stable for at least 1 year after receipt when stored at -20°C. Stock solutions are stable for up to 3 months at -20°C. Long Term Storage: -20°C
Catalog No. NC9852364