Learn More
StatLab™ Reserve Hematoxylin
SDP
Catalog No. STLSL200
Change view
Click to view available options
Quantity:
500 mL
Catalog No. Quantity
STLSL200 500 mL
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLSL200 Supplier StatLab™ Supplier No. SL200
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Uniquely formulated hematoxylin stain that can be used in place of Gill or Harris hematoxylins.
  • Does not require daily filtering or form metallic sheens
  • Rapidly provides unsurpassed nuclear chromatin delineation with no affinity for background or mucin staining
  • May be used for both histology and cytology applications
  • Reserve stains can last 3500 slides before losing intensity
Specifications
Quantity 500 mL
Product Type Hematoxylin Stain