Learn More
StatLab™ Reserve Multichrome Eosin
SDP
Catalog No. STLSL201
Change view
Click to view available options
Quantity:
500 mL
Catalog No. Quantity
STLSL201 500 mL
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLSL201 Supplier StatLab™ Supplier No. SL201
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Proprietary formulation that provides striking contrast and brilliant staining of all cytoplasmic components.
  • Three-dye combination provides familiar and vibrant cytoplasmic staining and differentiation
  • Alcoholic and acidified
  • Reserve stains can last 3500 slides before losing intensity
Specifications
Quantity 500 mL
Product Type Eosin Stain