Learn More
Andwin Scientific RESERVOIR 100ML STRL 200/CS
SDP
Catalog No. NC0272719
Encompass_Preferred
The disposable pipetting reservoirs are made for multichannel pipetting applications. This product is compatible with any type of pipettors that are up to 12-channel width. This polystyrene reservoir has a unique trough construction, graduation marks on interior walls, and pour-off spouts on each corner. It has a volume capacity of 100 mL and has a dimension of 14.6 x 8.3 x 3 cm. This product is sold in a case of 200 white sterile reservoirs.
Catalog No. NC0272719 Supplier Andwin Scientific Supplier No. S1005200
The disposable pipetting reservoirs are made for multichannel pipetting applications. This product is compatible with any type of pipettors that are up to 12-channel width. This polystyrene reservoir has a unique trough construction, graduation marks on interior walls, and pour-off spouts on each corner. It has a volume capacity of 100 mL and has a dimension of 14.6 x 8.3 x 3 cm. This product is sold in a case of 200 white sterile reservoirs.