Learn More
Agilent Technologies Reservoir, 12 column, 25/pk
Catalog No. NC1884483
Encompass
Reservoir, 12 column, polypropylene, 7 mL/column, 84 mL max, pyramid base geometries, 19 mm height, irradiated, 25/pk.
Catalog No. NC1884483 Supplier Agilent Technologies Supplier No. 204365100
Reservoir, 12 column, polypropylene, 7 mL/column, 84 mL max, pyramid base geometries, 19 mm height, irradiated, 25/pk.