Learn More
MTC Bio Reservoir, 25ml sterile, 5/ sterile bag
Catalog No. 501127244
Encompass_Preferred
This item has a minimum qty of 3 per supplier requirements.
MTC Bios multi-channel solution reservoirs are designed for use with all multichannel pipettes. Standard 4 channel, 8 channel, 12 channel and various special purpose pipettes are all compatible with these disposable, polystyrene reservoirs. To facilitate easy, dripless pour-off of unused reagents and solutions, all versions have indented spouts in each corner. Bottoms are formed with a sharply tapered V shape to minimize waste and reagent consumption. Whether your application calls for bulk packaging, packs of 5, or individually wrapped reservoirs, MTC reservoirs are supplied sterile and are packed in environmentally friendly packaging, within biodegradable bags and recycled cardboard boxes.
Catalog No. 50-112-7244 Supplier MTC Bio Supplier No. P80255S
This item has a minimum qty of 3 per supplier requirements.
MTC Bios multi-channel solution reservoirs are designed for use with all multichannel pipettes. Standard 4 channel, 8 channel, 12 channel and various special purpose pipettes are all compatible with these disposable, polystyrene reservoirs. To facilitate easy, dripless pour-off of unused reagents and solutions, all versions have indented spouts in each corner. Bottoms are formed with a sharply tapered V shape to minimize waste and reagent consumption. Whether your application calls for bulk packaging, packs of 5, or individually wrapped reservoirs, MTC reservoirs are supplied sterile and are packed in environmentally friendly packaging, within biodegradable bags and recycled cardboard boxes.