Learn More
Grainger Ergodyne Chill-Its Vest, Dry Evaporative, 2XL, Zipper Closure, Green, 1 ea
Catalog No. NC2392868
Encompass_Preferred
The Chill-Its 6685 Dry Evaporative Cooling Vest provides immediate cooling relief by utilizing dry evaporative technology to keep the wearer dry and comfortable. Designed for industrial applications, the vest features a lightweight construction that lasts up to 3 days and does not require refrigeration or gels for activation. The cooling material is treated with an anti-microbial agent to inhibit mold growth.
  • Dry evaporative cooling technology for long-lasting relief
  • Lightweight and machine washable
  • Features V-neck design with front zipper closure
  • Mesh side panels provide ventilation and stretch for a comfortable fit
  • Requires 17 oz (500 mL) of water to activate
  • Constructed with durable nylon
Catalog No. NC2392868 Supplier Grainger Supplier No. 40LR13
The Chill-Its 6685 Dry Evaporative Cooling Vest provides immediate cooling relief by utilizing dry evaporative technology to keep the wearer dry and comfortable. Designed for industrial applications, the vest features a lightweight construction that lasts up to 3 days and does not require refrigeration or gels for activation. The cooling material is treated with an anti-microbial agent to inhibit mold growth.
  • Dry evaporative cooling technology for long-lasting relief
  • Lightweight and machine washable
  • Features V-neck design with front zipper closure
  • Mesh side panels provide ventilation and stretch for a comfortable fit
  • Requires 17 oz (500 mL) of water to activate
  • Constructed with durable nylon