Learn More
Arcticwhite LLC 384 Well Reservoir, Half Height, Pyramid Bottom, Polypropylene (PP), 170 ML, 25 Reagent Reservoirs per Case
SDP
Catalog No. NC1853414
Encompass
384 Well Reservoir, Half Height, Pyramid Bottom, Polypropylene (PP), 170 ML, 25 Reagent Reservoirs per Case. This reservoir supports automated liquid handling and multichannel pipetting. Format - 384-well. Well shape - pyramid. Height - half-height. Capacity - 170 mL. Material - polypropylene (PP). Packaging - 25 per case. Supplied by Arctic White as 25 reagent reservoirs per case.
Catalog No. NC1853414 Supplier Arcticwhite LLC Supplier No. AWLSS30042
384 Well Reservoir, Half Height, Pyramid Bottom, Polypropylene (PP), 170 ML, 25 Reagent Reservoirs per Case. This reservoir supports automated liquid handling and multichannel pipetting. Format - 384-well. Well shape - pyramid. Height - half-height. Capacity - 170 mL. Material - polypropylene (PP). Packaging - 25 per case. Supplied by Arctic White as 25 reagent reservoirs per case.