Learn More
Electron Microscopy Sciences Disposible Reagent Reservoir, 50 ml, Sterile
SDP
Catalog No. 5033399
Encompass_Preferred
Unique product, "Trough within a trough" maximizes reagent recovery. Pour-off spouts on all four corners for convenience. A wide base provides stability and avoids spills. Graduated marks on inside wall for easy measuring. Sterilized by gamma irradiation.
Capacity: 50 ml
Sterile: Sterile
Catalog No. 50-333-99 Supplier Electron Microscopy Sciences Supplier No. 7049535
Unique product, "Trough within a trough" maximizes reagent recovery. Pour-off spouts on all four corners for convenience. A wide base provides stability and avoids spills. Graduated marks on inside wall for easy measuring. Sterilized by gamma irradiation.
Capacity: 50 ml
Sterile: Sterile