Learn More
Electron Microscopy Sciences Disposible Reagent Reservoir, 50 ml, Individually wrapped
SDP
Catalog No. 5033401
Encompass_Preferred
Unique product, "Trough within a trough" maximizes reagent recovery. Pour-off spouts on all four corners for convenience. A wide base provides stability and avoids spills. Graduated marks on inside wall for easy measuring.
Capacity: 50 ml
Sterile: Individually wrapped
Catalog No. 50-334-01 Supplier Electron Microscopy Sciences Supplier No. 7049635
Unique product, "Trough within a trough" maximizes reagent recovery. Pour-off spouts on all four corners for convenience. A wide base provides stability and avoids spills. Graduated marks on inside wall for easy measuring.
Capacity: 50 ml
Sterile: Individually wrapped