Learn More
Chemglass Life Sciences Reservoir, 55mL, Solution Basin, Clear PVC, Non-Sterile
SDP
Catalog No. 501532335
Encompass_Preferred
Reservoir, 55mL, Solution Basin, Clear PVC, Non-Sterile. PVC reservoirs, non-sterile, disposable, clear, 55mL capacity, for single or multi-channel pipettors. Material: Clear PVC. Angled sides and V-shaped bottom permits easy access to solution. Basin has transparent walls to permit easy viewing of contents. Can be used with single or multi-channel pipettors. Maximum capacity is 55mL
Catalog No. 50-153-2335 Supplier Chemglass Life Sciences Supplier No. CLS3797A100
Reservoir, 55mL, Solution Basin, Clear PVC, Non-Sterile. PVC reservoirs, non-sterile, disposable, clear, 55mL capacity, for single or multi-channel pipettors. Material: Clear PVC. Angled sides and V-shaped bottom permits easy access to solution. Basin has transparent walls to permit easy viewing of contents. Can be used with single or multi-channel pipettors. Maximum capacity is 55mL