Learn More
Chemglass Life Sciences 2000mL Reservoir, Chromatography, 75/50 Lower Ball
SDP
Catalog No. CG120037
Encompass_Preferred
2000mL Reservoir, Chromatography, 75/50 Lower Ball. Reservoir for use with CG-1199 and CG-1200 columns. Reservoir has a top 35/20 socket joint for use with CG-1201 flow control adapter and the listed size lower ball joint.
Catalog No. CG120037 Supplier Chemglass Life Sciences Supplier No. CG120037
2000mL Reservoir, Chromatography, 75/50 Lower Ball. Reservoir for use with CG-1199 and CG-1200 columns. Reservoir has a top 35/20 socket joint for use with CG-1201 flow control adapter and the listed size lower ball joint.