Learn More
Chemglass Life Sciences 1000mL Reservoir, Chromatography, 45/50 Lower Inner
SDP
Catalog No. 50873375
Encompass_Preferred
1000mL Reservoir, Chromatography, 45/50 Lower Inner. Reservoirs for use with CG-1188 and CG-1189 chromatography columns. Reservoir has a top 24/40 standard taper outer joint for use with CG-1191 flow control adapter and the listed size lower inner joint.
Catalog No. 50-873-375 Supplier Chemglass Life Sciences Supplier No. CG119014
1000mL Reservoir, Chromatography, 45/50 Lower Inner. Reservoirs for use with CG-1188 and CG-1189 chromatography columns. Reservoir has a top 24/40 standard taper outer joint for use with CG-1191 flow control adapter and the listed size lower inner joint.