Learn More
Ace Glass, Inc. Reservoir, solvent, plastic coated, 500mL, 3-valve Omnifit cap
SDP
Catalog No. 50978364
Encompass
500mL plastic-coated solvent reservoir is 14 psi rated, graduated, and features an Omnifit, 3-valve cap. Individually controlled valves permit the application of gas under pressure for venting, flushing, delivery, or pressure equalizing. Must be adequately shielded when under pressure. Filter accessories press directly on the caps 1/8 O.D. tubing providing filtration or sparging applications.
Catalog No. 50-978-364 Supplier Ace Glass, Inc. Supplier No. 5414139
500mL plastic-coated solvent reservoir is 14 psi rated, graduated, and features an Omnifit, 3-valve cap. Individually controlled valves permit the application of gas under pressure for venting, flushing, delivery, or pressure equalizing. Must be adequately shielded when under pressure. Filter accessories press directly on the caps 1/8 O.D. tubing providing filtration or sparging applications.