Learn More
Electron Microscopy Sciences Bench Top Reagent Reservoir Cooler, 100 ml volume
SDP
Catalog No. 50930356
Encompass_Preferred
Keeps reagents cool at ~5 degree C for approximately 3 hours without the mess of a cumbersome ice bath. Frozen gel is contained in a sealed molded cavity. Cooling extends safe working time for temperature sensitive reagents. Each 100 ml Reservoir fits perfectly and snuggly onto the Cooler ensuring uniform cooling.
Volume: 100 ml
Catalog No. 50-930-356 Supplier Electron Microscopy Sciences Supplier No. 7049750
Keeps reagents cool at ~5 degree C for approximately 3 hours without the mess of a cumbersome ice bath. Frozen gel is contained in a sealed molded cavity. Cooling extends safe working time for temperature sensitive reagents. Each 100 ml Reservoir fits perfectly and snuggly onto the Cooler ensuring uniform cooling.
Volume: 100 ml