Learn More
MilliporeSigma™ Supelco™ Reservoir for Filtration Apparatus 1
Catalog No. 111004719 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
11-100-4719 250 mL
11-100-4653 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-4719 Supplier MilliporeSigma™ Supelco™ Supplier No. 58063
Add to Cart
Add to Cart
Replacement glass reservoir for Supelco™ mobile phase filtration apparatus 1
Reservoir for Filtration Apparatus 1 protects the instrument and columns by removing particles and gases from solvents and other mobile phase components.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Liquid Chromatography
For Use With (Equipment) Filtration Apparatus 1
Volume (Metric) 250 mL