Learn More
Chemglass Life Sciences Chemglass Reservoir for 20 L Gas Scrubber (CG-1830-51), (2) 45/50 Side Necks, 100 mm Center Flange
SDP
Catalog No. 502733578
Encompass_Preferred
Chemglass CG-1830-51 reservoir is the lower collection and absorption vessel for a 20 L gas scrubber assembly. It is designed to integrate with glass columns and fittings using standardized taper joints and a center flange.
  • Holds 20 L volume.
  • Two 45/50 side necks for taper joint connections.
  • 100 mm center flange for column integration.
  • Intended for use as the reservoir component in 20 L gas scrubber systems.
  • Laboratory glass construction suitable for integration with standard glassware.
Catalog No. 50-273-3578 Supplier Chemglass Life Sciences Supplier No. CG183051
Chemglass CG-1830-51 reservoir is the lower collection and absorption vessel for a 20 L gas scrubber assembly. It is designed to integrate with glass columns and fittings using standardized taper joints and a center flange.
  • Holds 20 L volume.
  • Two 45/50 side necks for taper joint connections.
  • 100 mm center flange for column integration.
  • Intended for use as the reservoir component in 20 L gas scrubber systems.
  • Laboratory glass construction suitable for integration with standard glassware.