Learn More
Ace Glass, Inc. Reservoir, solvent, netted, 1000mL, 3-valve Omnifit cap
SDP
Catalog No. 50971869
Encompass
1000mL netted solvent reservoir is 14 psi rated, graduated, and features an Omnifit, 3-valve cap. Individually controlled valves permit the application of gas under pressure for venting, flushing, delivery, or pressure equalizing. Must be adequately shielded when under pressure. Filter accessories press directly on the caps 1/8 O.D. tubing providing filtration or sparging applications.
Catalog No. 50-971-869 Supplier Ace Glass, Inc. Supplier No. 541415
1000mL netted solvent reservoir is 14 psi rated, graduated, and features an Omnifit, 3-valve cap. Individually controlled valves permit the application of gas under pressure for venting, flushing, delivery, or pressure equalizing. Must be adequately shielded when under pressure. Filter accessories press directly on the caps 1/8 O.D. tubing providing filtration or sparging applications.