Learn More
Chemglass Life Sciences Reservoir White, Three Channel, Divided 25mL *3-Channels 5mL, Non-Sterile
SDP
Catalog No. 50143842
Encompass_Preferred
Reservoir White, Three Channel, Divided 25mL *3-Channels 5mL, Non-Sterile. Reservoirs are designed for use in multichannel pipetting applications. Our disposable reservoirs accommodate all types of pipettors up to 12-channel width. They feature a trough design, graduation marks on interior walls, and pour-off spouts on each corner. All sterile reservoirs are lot-certified, nonpyrogenic, and DNase- and RNase-free.The 25mL reservoirs are also available in a unique divided format that features either two or three distinct mini reservoir chambers. The two channel reservoir has 5mL and 10ml capacities. The three channel one has 5mL capacity each. All sizes are manufactured from virgin modified polystyrene. Available non-sterile or electron beam sterilized.
Catalog No. 50-143-842 Supplier Chemglass Life Sciences Supplier No. CLS3798019
Reservoir White, Three Channel, Divided 25mL *3-Channels 5mL, Non-Sterile. Reservoirs are designed for use in multichannel pipetting applications. Our disposable reservoirs accommodate all types of pipettors up to 12-channel width. They feature a trough design, graduation marks on interior walls, and pour-off spouts on each corner. All sterile reservoirs are lot-certified, nonpyrogenic, and DNase- and RNase-free.The 25mL reservoirs are also available in a unique divided format that features either two or three distinct mini reservoir chambers. The two channel reservoir has 5mL and 10ml capacities. The three channel one has 5mL capacity each. All sizes are manufactured from virgin modified polystyrene. Available non-sterile or electron beam sterilized.