Learn More
Beckman Coulter RESERVOIR STERILE 75ML 24/CS
Catalog No. NC9226465
Encompass
Half servoir (75mL) The Modular Reservoir System holds liquids for pipetting with Biomek Pipette Tools and Beckman Coulter Manual Pipettors
Catalog No. NC9226465 Supplier Beckman Coulter Supplier No. 372786
Half servoir (75mL) The Modular Reservoir System holds liquids for pipetting with Biomek Pipette Tools and Beckman Coulter Manual Pipettors