Learn More
Chemglass Life Sciences Reservoir Tray, Microplate, 85mL Reservoir, Low Profile, Flat Bottom, Polypropylene, Porvair, CASE OF 25
SDP
Catalog No. 501944005
Encompass_Preferred
Reservoir Tray, Microplate, 85mL Reservoir, Low Profile, Flat Bottom, Polypropylene, Porvair, CASE OF 25. Porvair reservoir trays are designed for use with any robotic liquid handling system. These reservoirs offer options for a single liquid or a partitioned space for several liquids. Most working configurations are accounted for as well as varying liquid volumes.
Catalog No. 50-194-4005 Supplier Chemglass Life Sciences Supplier No. CLS3590085
Reservoir Tray, Microplate, 85mL Reservoir, Low Profile, Flat Bottom, Polypropylene, Porvair, CASE OF 25. Porvair reservoir trays are designed for use with any robotic liquid handling system. These reservoirs offer options for a single liquid or a partitioned space for several liquids. Most working configurations are accounted for as well as varying liquid volumes.