Learn More
Chemglass Life Sciences Reservoir Tray, Microplate, 384 Well Reservoir 2 Control Wells, Pyramid Bottom, 282mL Polypropylene, Porvair, CASE OF 25
SDP
Catalog No. 501944006
Encompass_Preferred
Reservoir Tray, Microplate, 384 Well Reservoir 2 Control Wells, Pyramid Bottom, 282mL Polypropylene, Porvair, CASE OF 25. Porvair reservoir trays are designed for use with any robotic liquid handling system. These reservoirs offer options for a single liquid or a partitioned space for several liquids. Most working configurations are accounted for as well as varying liquid volumes.
Catalog No. 50-194-4006 Supplier Chemglass Life Sciences Supplier No. CLS3590384
Reservoir Tray, Microplate, 384 Well Reservoir 2 Control Wells, Pyramid Bottom, 282mL Polypropylene, Porvair, CASE OF 25. Porvair reservoir trays are designed for use with any robotic liquid handling system. These reservoirs offer options for a single liquid or a partitioned space for several liquids. Most working configurations are accounted for as well as varying liquid volumes.