Learn More
Chemglass Life Sciences Reservoir Tray, Microplate, 24 Column Reservoir/Each of 3.5mL, Low Profile, Partitioned, Pyramid Bottom, Polypropylene, Porvair, CASE OF 25
SDP
Catalog No. 501944007
Encompass_Preferred
Reservoir Tray, Microplate, 24 Column Reservoir/Each of 3.5mL, Low Profile, Partitioned, Pyramid Bottom, Polypropylene, Porvair, CASE OF 25. Porvair reservoir trays are designed for use with any robotic liquid handling system. These reservoirs offer options for a single liquid or a partitioned space for several liquids. Most working configurations are accounted for as well as varying liquid volumes.
Catalog No. 50-194-4007 Supplier Chemglass Life Sciences Supplier No. CLS3590024
Add to Cart
Add to Cart
Reservoir Tray, Microplate, 24 Column Reservoir/Each of 3.5mL, Low Profile, Partitioned, Pyramid Bottom, Polypropylene, Porvair, CASE OF 25. Porvair reservoir trays are designed for use with any robotic liquid handling system. These reservoirs offer options for a single liquid or a partitioned space for several liquids. Most working configurations are accounted for as well as varying liquid volumes.