Learn More
Electron Microscopy Sciences Disposible Reagent Reservoir with divider, 25 ml, Sterile
SDP
Catalog No. 501899331
Encompass_Preferred
Reservoir with divider with divider accommodates eight channel pipets on one side and one to four channel pipets on the other. The divider is also convenient for adding controls.
Capacity: 25 ml
Sterile: Sterile
Catalog No. 50-189-9331 Supplier Electron Microscopy Sciences Supplier No. 7049520
Reservoir with divider with divider accommodates eight channel pipets on one side and one to four channel pipets on the other. The divider is also convenient for adding controls.
Capacity: 25 ml
Sterile: Sterile