Learn More
Integra Biosciences Corp RESERVOIRS, 10ML, BULK 200/PK
SDP
Catalog No. NC0382499
Encompass
10mL disposable reagent reservoirs, sterile, four sleeves of 50 reservoirs per case INTEGRa reservoirs have been designed to nest inside each other, making it possible to get twice as many reservoirs in half the space of other products on the market, reducing inventory space requirements and shipping costs. Unlike traditional reservoirs that have hard to see graduations molded directly into the plastic, INTEGRa reservoirs are made of crystal clear polystyrene and fit into a reusable base with bold, crisp, clearly visible graduation markings. The unique design leads to more accurate reagent measurements, no over pouring, and less waste
Catalog No. NC0382499 Supplier Integra Biosciences Corp Supplier No. 4332
10mL disposable reagent reservoirs, sterile, four sleeves of 50 reservoirs per case INTEGRa reservoirs have been designed to nest inside each other, making it possible to get twice as many reservoirs in half the space of other products on the market, reducing inventory space requirements and shipping costs. Unlike traditional reservoirs that have hard to see graduations molded directly into the plastic, INTEGRa reservoirs are made of crystal clear polystyrene and fit into a reusable base with bold, crisp, clearly visible graduation markings. The unique design leads to more accurate reagent measurements, no over pouring, and less waste