Learn More
Electron Microscopy Sciences Disposible Reagent Reservoir, 25 ml, Non-sterile
SDP
Catalog No. 501899339
Encompass_Preferred
Unique product, "Trough within a trough" maximizes reagent recovery. Pour-off spouts on all four corners for convenience. A wide base provides stability and avoids spills. Graduated marks on inside wall for easy measuring.
Capacity: 25 ml
Sterile: Non-sterile
Catalog No. 50-189-9339 Supplier Electron Microscopy Sciences Supplier No. 7049730
Unique product, "Trough within a trough" maximizes reagent recovery. Pour-off spouts on all four corners for convenience. A wide base provides stability and avoids spills. Graduated marks on inside wall for easy measuring.
Capacity: 25 ml
Sterile: Non-sterile