Learn More
Med Vet International Merck, Resflor Gold, 300mg/mL, 250mL
Catalog No. 502828596
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
An injectable solution of the synthetic antibiotic florfenicol (300 mg/mL) and the non-steroidal anti-inflammatory drug (NSAID) flunixin meglumine (16.5 mg/mL). Also contains 2-pyrrolidone 300 mg/mL, malic acid 35 mg/mL, and triacetin q.s. It is indicated for treatment of bovine respiratory disease (BRD) associated with Mannheimia haemolytica, Pasteurella multocida, and Histophilus somni, and control of BRD-associated pyrexia in beef and non-lactating dairy cattle. Do not use in female dairy cattle 20 months or older, or in calves to be processed for veal.
Catalog No. 50-282-8596 Supplier Med Vet International Supplier No. RXRESFLOR250ML
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
An injectable solution of the synthetic antibiotic florfenicol (300 mg/mL) and the non-steroidal anti-inflammatory drug (NSAID) flunixin meglumine (16.5 mg/mL). Also contains 2-pyrrolidone 300 mg/mL, malic acid 35 mg/mL, and triacetin q.s. It is indicated for treatment of bovine respiratory disease (BRD) associated with Mannheimia haemolytica, Pasteurella multocida, and Histophilus somni, and control of BRD-associated pyrexia in beef and non-lactating dairy cattle. Do not use in female dairy cattle 20 months or older, or in calves to be processed for veal.