Learn More
Med Vet International Merck, Resflor Gold, 300mg/mL, 500mL
Catalog No. 502828597
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
(Florfenicol and Flunixin Meglumine) A simple bovine respiratory disease (BRD) treatment strategy. It involves a combination of two therapies in one dose: The powerful antibiotic florfenicol to kill or inhibit the disease-causing bacteria mannheimia haemolytica, pasteurella multocida, histoophilus somni, and mycoplama bovis. The fast-acting non-steroidal anti-inflammatory drug (NSAID) flunixin meglumine to reduce BRD-associated fever.
Catalog No. 50-282-8597 Supplier Med Vet International Supplier No. RXRESFLOR500ML
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
(Florfenicol and Flunixin Meglumine) A simple bovine respiratory disease (BRD) treatment strategy. It involves a combination of two therapies in one dose: The powerful antibiotic florfenicol to kill or inhibit the disease-causing bacteria mannheimia haemolytica, pasteurella multocida, histoophilus somni, and mycoplama bovis. The fast-acting non-steroidal anti-inflammatory drug (NSAID) flunixin meglumine to reduce BRD-associated fever.