Learn More
Electron Microscopy Sciences DiATOME Histo Jumbo Diamond Knife 6.0 mm, 45° Knife angle
SDP
Catalog No. 501898865
Encompass
A resharpened DiATOME knife has the same length, an identical cutting edge and carries the same guarantee of quality as a new knife. During resharpening a knife may be reworked into another type.
Size: 6.0 mm
Knife angle: 45°
Catalog No. 50-189-8865 Supplier Electron Microscopy Sciences Supplier No. 60HISJR
A resharpened DiATOME knife has the same length, an identical cutting edge and carries the same guarantee of quality as a new knife. During resharpening a knife may be reworked into another type.
Size: 6.0 mm
Knife angle: 45°