Learn More
Electron Microscopy Sciences DiATOME Histo Jumbo Diamond Knife 8.0 mm, 45° Knife angle
SDP
Catalog No. 501900065
Encompass
A resharpened DiATOME knife has the same length, an identical cutting edge and carries the same guarantee of quality as a new knife. During resharpening a knife may be reworked into another type.
Size: 8.0 mm
Knife angle: 45°
Catalog No. 50-190-0065 Supplier Electron Microscopy Sciences Supplier No. 80HISJR
A resharpened DiATOME knife has the same length, an identical cutting edge and carries the same guarantee of quality as a new knife. During resharpening a knife may be reworked into another type.
Size: 8.0 mm
Knife angle: 45°