Learn More
Cayman Chemical Resibufogenin, Purity: ≥98%, CAS: 465-39-4, 10 mg
SDP
Catalog No. 502893964
Encompass_Preferred
A cardiac glycoside that inhibits the Na+/K+-ATPase; exhibits cardiotonic, vasopressor, and respiratory stimulating actions in both animal and clinical models
Catalog No. 50-289-3964 Supplier Cayman Chemical Supplier No. 900134810 MG
A cardiac glycoside that inhibits the Na+/K+-ATPase; exhibits cardiotonic, vasopressor, and respiratory stimulating actions in both animal and clinical models