Learn More
Enzo Life Sciences Resibufogenin (10mg). CAS: 465-39-4
SDP
Supplier:  Enzo Life Sciences ALX350286M010
Encompass_Preferred
Specific Na+/K+-ATPase inhibitor. Alternative name: (3beta,5beta,15beta)-14,15Epoxy-3-hydroxy-5-bufa-20,22-dienolide. Purity: ≥98% (HPLC). Solubility: Soluble in chloroform. Long Term Storage: +4°C.
Catalog No. 50-200-8829