Learn More
Cayman Chemical ResIbufogenIn 1mg
SDP
Catalog No. 502897021
Encompass_Preferred
A cardiac glycoside that inhibits the Na+/K+-ATPase; exhibits cardiotonic, vasopressor, and respiratory stimulating actions in both animal and clinical models
Catalog No. 50-289-7021 Supplier Cayman Chemical Supplier No. 90013481 MG
A cardiac glycoside that inhibits the Na+/K+-ATPase; exhibits cardiotonic, vasopressor, and respiratory stimulating actions in both animal and clinical models