Learn More
TARGETMOL CHEMICALS INC RESIBUFOGENIN 5MG
SDP
Catalog No. 502579839
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Resibufogenin (Bufogenin) is a cytotoxic steroid isolated from the Chinese drug ChanSu. 2. Resibufogenin can inhibit rectifier potassium current ( I K ) and transient potassium current ( I A ) it has pathological effects on central nervous system. 3. Resibufogenin corrects hypertension in a rat model of human preeclampsia it not only prevents the advent of hypertension and proteinuria but also the development of intrauterine growth restriction.4. Resibufogenina's major metabolic pathway in the human liver is CYP3A4 dominantly mediated 5(beta)-hydroxylation . purity: 98%
Catalog No. 50-257-9839 Supplier TARGETMOL CHEMICALS INC Supplier No. T4A24585MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Resibufogenin (Bufogenin) is a cytotoxic steroid isolated from the Chinese drug ChanSu. 2. Resibufogenin can inhibit rectifier potassium current ( I K ) and transient potassium current ( I A ) it has pathological effects on central nervous system. 3. Resibufogenin corrects hypertension in a rat model of human preeclampsia it not only prevents the advent of hypertension and proteinuria but also the development of intrauterine growth restriction.4. Resibufogenina's major metabolic pathway in the human liver is CYP3A4 dominantly mediated 5(beta)-hydroxylation . purity: 98%