Learn More
Cygnus Technologies CHO Host Cell DNA Detection Kit in Tubes
Catalog No. NC0791049
Encompass
Used to measure residual CHO host cell DNA, this kit uses a 2 ml microfuge tube format and includes our proprietary extraction procedure, which does not use iodine or glycogen. The superior co-precipitating reagents do not interfere with qPCR and ensure that pellets do not become dislodged during extraction. This eliminates the need to centrifiuge samples or repeat a run. Cell line-specific, CHO DNA calibrators are included in the kit. Upon completion of extraction, samples are reacted with PicoGreen solution. Time to completion is under 2 hours. Each kit supplies enough reagent for the analysis of 50 samples in duplicate.
Catalog No. NC0791049 Supplier Cygnus Technologies Supplier No. D550T
Used to measure residual CHO host cell DNA, this kit uses a 2 ml microfuge tube format and includes our proprietary extraction procedure, which does not use iodine or glycogen. The superior co-precipitating reagents do not interfere with qPCR and ensure that pellets do not become dislodged during extraction. This eliminates the need to centrifiuge samples or repeat a run. Cell line-specific, CHO DNA calibrators are included in the kit. Upon completion of extraction, samples are reacted with PicoGreen solution. Time to completion is under 2 hours. Each kit supplies enough reagent for the analysis of 50 samples in duplicate.