Learn More
American Research Products Inc Residual Protein A Kit
Catalog No. 5000801919
Encompass
Universal Residual Protein A Detection ELISA Kit; Residual Detection of Various Types of Protein A in Antibodies, Fc-fusion Proteins and other Products
Catalog No. 50-008-01919 Supplier American Research Products Inc Supplier No. ARPERPA01
Universal Residual Protein A Detection ELISA Kit; Residual Detection of Various Types of Protein A in Antibodies, Fc-fusion Proteins and other Products