Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Residual Solvent - Toluene Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111017858 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
3 x 1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7858 3 x 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7858 Supplier MilliporeSigma Supelco Supplier No. PHR13173X1.2ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 997°F
CAS 108-88-3
Vapor Pressure 26 mmHg (25°C)
Vapor Density 3.2 (vs. air)
Density 0.865 g/mL (at 25°C)
Flash Point 4.0°C
Linear Formula C6H5 CH3
MDL Number MFCD00008512
Melting Point -93°C
Boiling Point 110°C to 111°C
Synonym Toluene
Type Certified Reference Material
Quantity 3 x 1.2 mL
Formula Weight 92.14
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.