Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Residual Solvents, California Set 1, MilliporeSigma™ Supelco™

Catalog No. 111004438 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-4438 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-4438 Supplier MilliporeSigma Supelco Supplier No. CAN10031.2ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydrocarbons are often used in the production of extracts in the cannabis industry. Care should be taken to minimize residual solvents remaining on material intended for human consumption. To support this, many states have enacted regulations prescribing the routine analytical testing of residual solvents in cannabis-derived products. This multi-analyte Certified Reference Material is designed specifically to satisfy the needs of analytical testing in California.Five components have been carefully selected for convenience of use, while promoting long-term stability in solution. The analytes are formulated at 100 mug/g each, and are designed for a 100x dilution factor relative to the California action limits.For complete calibration of the required California residual solvents, this CRM may be used in conjunction with the other products in the series: CAN1008CAN1012CAN1013
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Flash Point 87°C
Type Certified Reference Material
Quantity 1.2 mL
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.