Learn More
SPEX CERTIPREP LLC 3P NPW-Residue, 500mL
Catalog No. NC9720605
Encompass_Preferred
(NSI by Zeptometrix) A 500 mL ready-to-use whole volume sample to be analyzed for Total Suspended Solids in the NELAC range of 20-100 mg/L and Total Solids formulated in the NELAC range of 140-800 mg/L.
Catalog No. NC9720605 Supplier SPEX CERTIPREP LLC 3P Supplier No. PEI079
(NSI by Zeptometrix) A 500 mL ready-to-use whole volume sample to be analyzed for Total Suspended Solids in the NELAC range of 20-100 mg/L and Total Solids formulated in the NELAC range of 140-800 mg/L.