The 3M Safety-Walk Slip-Resistant Medium Resilient Tapes and Treads 300 Series is a resilient, non-mineral, slip-resistant material constructed from a dimensionally stable plastic film with a resilient textured surface, bonded to a pressure-sensitive adhesive. It is designed for industrial/occupational use, providing a textured resilient high friction surface for long-lasting performance.
- Non-mineral, medium textured, slip-resistant vinyl surface
- Durable adhesive for long-lasting performance
- Soft textured for barefoot comfort and light shoe traffic
- NFSI certified 'slip resistant'
- Film technology allows repositioning before adhesion
- Adhesive technology ensures bubble-free application
- Suitable for flat, smooth indoor and outdoor surfaces
- Minimum application temperature: 10°C (50°F)
- Maximum service temperature: 79°C (175°F)