Learn More
MSC RESILIENT ANTI-SLIP TAPE
Catalog No. 5000686834
Encompass
This Floor & Aisle Marking Tape is 6 in. wide and 60 ft. long, constructed from vinyl, and features a gray, anti-slip surface.
  • Non-mineral, medium textured, slip-resistant vinyl surface
  • Textured resilient high friction surface and durable adhesive provide long-lasting performance
  • Suitable for barefoot- or shoe-traffic areas such as locker rooms, poolsides, boats, and exercise equipment
  • NFSI High Traction Certification
Catalog No. 50-006-86834 Supplier MSC Supplier No. 51556983
This Floor & Aisle Marking Tape is 6 in. wide and 60 ft. long, constructed from vinyl, and features a gray, anti-slip surface.
  • Non-mineral, medium textured, slip-resistant vinyl surface
  • Textured resilient high friction surface and durable adhesive provide long-lasting performance
  • Suitable for barefoot- or shoe-traffic areas such as locker rooms, poolsides, boats, and exercise equipment
  • NFSI High Traction Certification