Learn More
Bio Rad Laboratories RESIN AG 1-X2
Catalog No. 509013131
Encompass
Media, Ion exchange; Bio-Rad; Analytical Grade; AG 1-X2 resin; 500g; Acetate; 200-400 dry mesh; 75-180um wet bead
Catalog No. 50-901-3131 Supplier Bio Rad Laboratories Supplier No. 1401253
Media, Ion exchange; Bio-Rad; Analytical Grade; AG 1-X2 resin; 500g; Acetate; 200-400 dry mesh; 75-180um wet bead