Learn More
Bio Rad Laboratories RESIN AG 1-X8
Catalog No. 509012927
Encompass
Media, Biotechnology grade; Bio-Rad; AG 1-X8 resin; 100g; Hydroxide; 100-200 dry mesh; 106-180um wet bead
Catalog No. 50-901-2927 Supplier Bio Rad Laboratories Supplier No. 1432445
Media, Biotechnology grade; Bio-Rad; AG 1-X8 resin; 100g; Hydroxide; 100-200 dry mesh; 106-180um wet bead