Learn More
Bio Rad Laboratories RESIN AG 1-X8
Supplier:  Bio Rad Laboratories 1401431
Encompass
Media, Ion exchange; Bio-Rad; Analytical Grade; AG 1-X8 resin; 500g; Chloride; 50-100 dry mesh; 180-425um wet bead
Catalog No. 50-901-3112