Learn More
Bio Rad Laboratories RESIN AG 501-X8(D)
Catalog No. 509013155
Encompass
Media, Biotechnology grade; Bio-Rad; AG 501-X8(D) resin; 100g; Self-adsorbed; 50-100 dry mesh; 180-425um wet bead
Catalog No. 50-901-3155 Supplier Bio Rad Laboratories Supplier No. 1437425
Media, Biotechnology grade; Bio-Rad; AG 501-X8(D) resin; 100g; Self-adsorbed; 50-100 dry mesh; 180-425um wet bead