Learn More
Bio Rad Laboratories RESIN AG 50W-X2
Catalog No. 509012930
Encompass
Media, Biotechnology grade; Bio-Rad; AG 50W-X2 resin; 100g; Hydrogen; 100-200 dry mesh; 106-300um wet bead
Catalog No. 50-901-2930 Supplier Bio Rad Laboratories Supplier No. 1435241
Media, Biotechnology grade; Bio-Rad; AG 50W-X2 resin; 100g; Hydrogen; 100-200 dry mesh; 106-300um wet bead