Learn More
Bio Rad Laboratories RESIN AG 50W-X8
Catalog No. 509013124
Encompass
Media, Ion exchange; Bio-Rad; Analytical Grade; AG 50W-X8 resin; 500g; Hydrogen; 20-50 dry mesh; 300-1180um wet bead
Catalog No. 50-901-3124 Supplier Bio Rad Laboratories Supplier No. 1421421
Media, Ion exchange; Bio-Rad; Analytical Grade; AG 50W-X8 resin; 500g; Hydrogen; 20-50 dry mesh; 300-1180um wet bead