Learn More
Bio Rad Laboratories RESIN BIO-REX 5
Catalog No. 509010072
Encompass
Resin; Bio-Rad; Bio-Rex 5; Chloride; 100-200 dry mesh; 75-150um wet bead; 500g
Catalog No. 50-901-0072 Supplier Bio Rad Laboratories Supplier No. 1407841
Resin; Bio-Rad; Bio-Rex 5; Chloride; 100-200 dry mesh; 75-150um wet bead; 500g